(5-((4-(4-Chlorophenyl)piperidin-1-yl)sulfonyl)picolinoyl)glycine ≥98%

Supplier: Ambeed
Warning

Synonyms: USP5-IN-1

A1601387-1MG A1601387-10MG A1601387-5MG
77598-094EA 76.88 USD
77598-094 77625-536 77625-538
(5-((4-(4-Chlorophenyl)piperidin-1-yl)sulfonyl)picolinoyl)glycine ≥98%
(5-((4-(4-Chlorophenyl)piperidin-1-yl)sulfonyl)picolinoyl)glycine

Precaución: for Research Use Only

Formula: C₁₉H₂₀ClN₃O₅S
MW: 437.9 g/mol
MDL Number: MFCD35061808
CAS Number: 2851040-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR