1-((S)-4-(7-((R)-6-Amino-4-methyl-3-(trifluoromethyl)pyridin-2-yl)-6-chloro-8-fluoro-2-(((S)-1-methylpyrrolidin-2-yl)methoxy)quinazolin-4-yl)-3-methylpiperazin-1-yl)prop-2-en-1-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Divarasib

A1600972-1MG A1600972-10MG A1600972-5MG A1600972-100MG A1600972-25MG
77598-092EA 80.98 USD
77598-092 77625-528 77625-530 77876-352 77876-353
1-((S)-4-(7-((R)-6-Amino-4-methyl-3-(trifluoromethyl)pyridin-2-yl)-6-chloro-8-fluoro-2-(((S)-1-methylpyrrolidin-2-yl)methoxy)quinazolin-4-yl)-3-methylpiperazin-1-yl)prop-2-en-1-one ≥98%
1-((S)-4-(7-((R)-6-Amino-4-methyl-3-(trifluoromethyl)pyridin-2-yl)-6-chloro-8-fluoro-2-(((S)-1-methylpyrrolidin-2-yl)methoxy)quinazolin-4-yl)-3-methylpiperazin-1-yl)prop-2-en-1-one

Precaución: for Research Use Only

Formula: C₂₉H₃₂ClF₄N₇O₂
MW: 622.06 g/mol
CAS Number: 2417987-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR