Methyl 3-(2-bromoacetyl)benzoate ≥98%

Supplier: Ambeed
Danger

A160041-100MG A160041-5G
77683-332EA 38 USD
77683-332 77683-333
Methyl 3-(2-bromoacetyl)benzoate ≥98%
Methyl 3-(2-bromoacetyl)benzoate

Precaución: For research use only.

Formula: C₁₀H₉BrO₃
MW: 257.081 g/mol
MDL Number: MFCD09834617
CAS Number: 27475-19-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR