(R)-1,12-Bis(diphenylphosphanyl)-6,7-diphenyldibenzo[e,g][1,4]diazocine ≥98%

Supplier: Ambeed
Danger

A1597854-100MG A1597854-250MG
77625-506EA 321.36 USD
77625-506 77625-508
(R)-1,12-Bis(diphenylphosphanyl)-6,7-diphenyldibenzo[e,g][1,4]diazocine ≥98%
(R)-1,12-Bis(diphenylphosphanyl)-6,7-diphenyldibenzo[e,g][1,4]diazocine

Precaución: for Research Use Only

Formula: C₅₀H₃₆N₂P₂
MW: 726.78 g/mol
CAS Number: 2648055-15-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR