10-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-10-oxodecanoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: VH 032 amide-alkylC8-acid

A1595813-100MG A1595813-250MG A1595813-50MG A1595813-1G
77598-082EA 186.55 USD
77598-082 77598-084 77598-086 77625-492
10-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-10-oxodecanoic acid ≥98%
10-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-10-oxodecanoic acid

Precaución: for Research Use Only

Formula: C₃₂H₄₆N₄O₆S
MW: 614.8 g/mol
CAS Number: 2172819-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR