5-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-5-oxopentanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: VH 032 amide-alkylC3-acid

A1595811-100MG A1595811-250MG
77625-488EA 195.7 USD
77625-488 77625-490
5-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-5-oxopentanoic acid ≥95%
5-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-5-oxopentanoic acid

Precaución: for Research Use Only

Formula: C₂₇H₃₆N₄O₆S
MW: 544.66 g/mol
MDL Number: MFCD32903592
CAS Number: 2172819-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR