4-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-4-oxobutanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (S,R,S)-AHPC--C2-COOH

A1595810-250MG A1595810-50MG
77625-484EA 459.38 USD
77625-484 77625-486
4-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-4-oxobutanoic acid ≥95%
4-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₂₆H₃₄N₄O₆S
MW: 530.64 g/mol
MDL Number: MFCD32898802
CAS Number: 2172819-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR