5-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)pentanoic acid ≥95%

Supplier: Ambeed
Warning

A1595748-250MG A1595748-50MG A1595748-1G
77625-478EA 309 USD
77625-478 77625-480 77876-326
5-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)pentanoic acid ≥95%
5-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)pentanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₉N₃O₆
MDL Number: MFCD32063490
CAS Number: 2225940-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR