Ansamitocin P-3 98% P-3 major, mix(P-0, P-1, P-2, P-3, P-3’, P-4, P-4’)

Supplier: Ambeed
Warning

Synonyms: 3-O-de[2-(acetylmethylamino)-1-oxopropyl]-3-O-(1-oxobutyl)-maytansine, Maytansinol isobutyrate, Antibiotic C 15003P3, (14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-Cchloro-14-hydroxy-85,14-dimethoxy-33,2,7,10-tetramethyl-12,6-dioxo-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-dien-4-yl isobutyrate, NSC292222

A159572-1MG A159572-10MG A159572-5MG
77615-296EA 5.15 USD
77615-296 77746-089 77746-090
Ansamitocin P-3 98% P-3 major, mix(P-0, P-1, P-2, P-3, P-3’, P-4, P-4’)
Ansamitocin P-3

Precaución: for Research Use Only

Formula: C₃₂H₄₃ClN₂O₉
MW: 635.14 g/mol
MDL Number: MFCD00274586
CAS Number: 66584-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR