Methyl-5-methylthiazole-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A159566-1G A159566-250MG A159566-5G
77611-540EA 17.51 USD
77611-540 77883-251 77883-252
Methyl-5-methylthiazole-2-carboxylate ≥98%
Methyl-5-methylthiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MDL Number: MFCD01924773
CAS Number: 79247-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR