1-(6-Aminonaphthalen-2-yl)ethanone ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A158501-100MG A158501-250MG A158501-1G
77683-322EA 98 USD
77683-322 77683-323 77890-038
1-(6-Aminonaphthalen-2-yl)ethanone ≥95%, Ambeed.Inc
1-(6-Aminonaphthalen-2-yl)ethanone

Precaución: For research use only.

Formula: C₁₂H₁₁NO
MDL Number: MFCD20693622
CAS Number: 7470-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR