(8S,10S)-6,8,11-Trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-10-(((1S,3R,4aS,9S,9aR,10aS)-9-methoxy-1-methyloctahydro-1H-pyrano[4',3':4,5]oxazolo[2,3-c][1,4]oxazin-3-yl)oxy)-7,8,9,10-tetrahydrotetracene-5,12-dione ≥98%

Supplier: Ambeed
Warning

Synonyms: PNU-159682

A158340-10MG A158340-1MG A158340-5MG
77611-520EA 81.37 USD
77611-520 77611-522 77611-524
(8S,10S)-6,8,11-Trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-10-(((1S,3R,4aS,9S,9aR,10aS)-9-methoxy-1-methyloctahydro-1H-pyrano[4',3':4,5]oxazolo[2,3-c][1,4]oxazin-3-yl)oxy)-7,8,9,10-tetrahydrotetracene-5,12-dione ≥98%
(8S,10S)-6,8,11-Trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-10-(((1S,3R,4aS,9S,9aR,10aS)-9-methoxy-1-methyloctahydro-1H-pyrano[4',3':4,5]oxazolo[2,3-c][1,4]oxazin-3-yl)oxy)-7,8,9,10-tetrahydrotetracene-5,12-dione

Precaución: for Research Use Only

Formula: C₃₂H₃₅NO₁₃
MW: 641.62 g/mol
MDL Number: MFCD12756329
CAS Number: 202350-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR