2-(2,6-Dioxopiperidin-3-yl)-5-(piperazin-1-yl)isoindoline-1,3-dione hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: Thalidomide-piperazine HCl

A1581459-250MG A1581459-100MG
77625-466EA 420.24 USD
77625-466 77876-322
2-(2,6-Dioxopiperidin-3-yl)-5-(piperazin-1-yl)isoindoline-1,3-dione hydrochloride ≥98%
2-(2,6-Dioxopiperidin-3-yl)-5-(piperazin-1-yl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₉ClN₄O₄
MW: 378.81 g/mol
MDL Number: MFCD33404953
CAS Number: 2228029-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR