(2-Bromoethyl)cyclopropane ≥98%

Supplier: Ambeed
Danger

A158137-1G A158137-250MG
77883-244EA 278 USD
77883-244 77883-245
(2-Bromoethyl)cyclopropane ≥98%
(2-Bromoethyl)cyclopropane

Precaución: for Research Use Only

Formula: C₅H₉Br
MDL Number: MFCD16860592
CAS Number: 36982-56-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR