1-(3-Bromo-5-methoxyphenyl)ethanone ≥95%

Supplier: Ambeed
Warning

A157873-1G A157873-250MG
77611-512EA 176.13 USD
77611-512 77683-318
1-(3-Bromo-5-methoxyphenyl)ethanone ≥95%
1-(3-Bromo-5-methoxyphenyl)ethanone

Precaución: For research use only.

Formula: C₉H₉BrO₂
MW: 229.071 g/mol
MDL Number: MFCD22544400
CAS Number: 1073642-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR