(2-Methoxy-3-nitrophenyl)boronic acid ≥98%
Supplier: Ambeed
|
Warning
|
A1574988-100MG
A1574988-1G
77876-317EA
47
USD
77876-317
77876-318
(2-Methoxy-3-nitrophenyl)boronic acid ≥98%
(2-Methoxy-3-nitrophenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₇H₈BNO₅ |
MDL Number:
MFCD30735843 CAS Number: 2401013-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...