4-Bromo-2,5-dimethyl-2H-1,2,3-triazole ≥97%

Supplier: Ambeed
Warning

A157464-100MG A157464-1G A157464-250MG
77871-784EA 73 USD
77871-784 77871-785 77871-786
4-Bromo-2,5-dimethyl-2H-1,2,3-triazole ≥97%
4-Bromo-2,5-dimethyl-2H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₄H₆BrN₃
MDL Number: MFCD14706834
CAS Number: 942060-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR