Methyl 2-(2-bromophenyl)-2-oxoacetate ≥97%

Supplier: Ambeed
Warning

A157327-1G A157327-250MG
77683-314EA 56 USD
77683-314 77683-315
Methyl 2-(2-bromophenyl)-2-oxoacetate ≥97%
Methyl 2-(2-bromophenyl)-2-oxoacetate

Precaución: For research use only.

Formula: C₉H₇BrO₃
MW: 243.05 g/mol
MDL Number: MFCD18206246
CAS Number: 122394-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR