5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]thiazol-2-amine ≥98%

Supplier: Ambeed
Warning

A1569337-1G A1569337-250MG
77625-452EA 161.71 USD
77625-452 77625-454
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]thiazol-2-amine ≥98%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₂S
MW: 276.16 g/mol
MDL Number: MFCD16995821
CAS Number: 2043401-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR