Boc-3,5-diiodo-Tyr-OMe ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(4-hydroxy-3,5-diiodophenyl)propanoate

A156908-1G A156908-25G A156908-5G
77625-448EA 15.45 USD
77625-448 77877-039 77877-040
Boc-3,5-diiodo-Tyr-OMe ≥98%
Boc-3,5-diiodo-Tyr-OMe

Precaución: for Research Use Only

Formula: C₁₅H₁₉I₂NO₅
MW: 547.13 g/mol
MDL Number: MFCD00237523
CAS Number: 128781-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR