tert-Butyl but-3-yn-1-yl(methyl)carbamate ≥98%

Supplier: Ambeed
Warning

A1568829-1G A1568829-100MG A1568829-250MG
77598-072EA 524.8 USD
77598-072 77876-304 77876-305
tert-Butyl but-3-yn-1-yl(methyl)carbamate ≥98%
tert-Butyl but-3-yn-1-yl(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₂
MDL Number: MFCD22548359
CAS Number: 159492-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR