2,4-Dichloro-1,5-naphthyridine ≥95%

Supplier: Ambeed
Warning

A156853-1G A156853-250MG
77671-214EA 1004 USD
77671-214 77883-235
2,4-Dichloro-1,5-naphthyridine ≥95%
2,4-Dichloro-1,5-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
MDL Number: MFCD12828139
CAS Number: 28252-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR