5-Iodo-1-methylimidazole ≥98%

Supplier: Ambeed
Warning

A156638-1G A156638-5G A156638-250MG A156638-100MG A156638-10G
77611-498EA 39.14 USD
77611-498 77611-500 77671-208 77883-233 77883-234
5-Iodo-1-methylimidazole ≥98%
5-Iodo-1-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₅IN₂
MDL Number: MFCD01632215
CAS Number: 71759-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR