(2-Ethoxy-4,5-difluorophenyl)boronic acid ≥98%

Supplier: Ambeed

A156599-1G A156599-5G
77671-206EA 43 USD
77671-206 77883-232
(2-Ethoxy-4,5-difluorophenyl)boronic acid ≥98%
(2-Ethoxy-4,5-difluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BF₂O₃
MDL Number: MFCD07784347
CAS Number: 870778-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR