Methyl 2-fluoro-3-methoxybenzoate ≥97%

Supplier: Ambeed
Warning

A156498-1G A156498-5G
77890-026EA 12 USD
77890-026 77890-027
Methyl 2-fluoro-3-methoxybenzoate ≥97%
Methyl 2-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
MDL Number: MFCD14698048
CAS Number: 958991-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR