3-Chloro-5-(2,2,2-trifluoroethoxy)pyridazine ≥95%

Supplier: Ambeed
Warning

A156493-100MG A156493-1G A156493-250MG
77611-496EA 26.78 USD
77611-496 77683-309 77683-310
3-Chloro-5-(2,2,2-trifluoroethoxy)pyridazine ≥95%
3-Chloro-5-(2,2,2-trifluoroethoxy)pyridazine

Precaución: For research use only.

Formula: C₆H₄ClF₃N₂O
MW: 212.56 g/mol
MDL Number: MFCD19688849
CAS Number: 1346691-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR