(R)-2-Methyl-1-phenylpropan-1-amine hydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
A156237-1G
A156237-250MG
A156237-5G
77883-227EA
191
USD
77883-227
77883-228
77883-229
(R)-2-Methyl-1-phenylpropan-1-amine hydrochloride ≥95%
(R)-2-Methyl-1-phenylpropan-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₆ClN |
MDL Number:
MFCD12068520 CAS Number: 51600-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...