(Benzhydrylthio)acetic acid ≥97%

Supplier: Ambeed
Warning

A156217-100G A156217-500G
77667-204EA 31 USD
77667-204 77871-778
(Benzhydrylthio)acetic acid ≥97%
(Benzhydrylthio)acetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂S
MDL Number: MFCD05262040
CAS Number: 63547-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR