5-Bromo-1-methyl-1H-1,2,3-triazole ≥95%

Supplier: Ambeed
Warning

A156207-250MG A156207-100MG A156207-1G
77592-920EA 188.6 USD
77592-920 77883-225 77883-226
5-Bromo-1-methyl-1H-1,2,3-triazole ≥95%
5-Bromo-1-methyl-1H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₃H₄BrN₃
MDL Number: MFCD13190021
CAS Number: 16681-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR