Ethyl 3-bromopropiolate ≥96%

Supplier: Ambeed
Warning

A155595-100MG A155595-1G A155595-250MG A155595-5G
77890-018EA 47 USD
77890-018 77890-019 77890-020 77890-021
Ethyl 3-bromopropiolate ≥96%
Ethyl 3-bromopropiolate

Precaución: for Research Use Only

Formula: C₅H₅BrO₂
MDL Number: MFCD11977143
CAS Number: 41658-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR