Copper(II) dimethyldithiocarbamate 98

Supplier: Ambeed
Danger

Synonyms: Copperdimethyldithiocarbamate, Dimethyldithiocarbamic Acid Copper(II) Salt

A155532-5G A155532-25G A155532-100G A155532-500G
77600-224EA 52.28 USD
77600-224 77625-420 77625-418 77879-074
Copper(II) dimethyldithiocarbamate 98
Copper(II) dimethyldithiocarbamate

Precaución: for Research Use Only

Formula: C₃H₇NS₂
MW: 303.96 g/mol
MDL Number: MFCD00050845
CAS Number: 137-29-1
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR