Clarithromycin ≥98%

Supplier: Ambeed
Warning

Synonyms: (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-(((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-12,13-dihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-7-methoxy-3,5,7,9,11,13-hexamethyloxacyclotetradecane-2,10-dione

A155509-5G A155509-25G A155509-100G
77600-222EA 51.25 USD
77600-222 77600-220 77625-416
Clarithromycin ≥98%
Clarithromycin

Precaución: for Research Use Only

Formula: C₃₈H₆₉NO₁₃
MW: 747.96 g/mol
MDL Number: MFCD00865140
CAS Number: 81103-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR