Butyl (S)-(-)-lactate ≥97%

Supplier: Ambeed
Warning

Synonyms: (S)-Butyl 2-hydroxypropanoate, n-Butyl L-lactate, (-)-Butyl L-lactate, (S)-(-)-Butyl lactate, Butyl (S)-(-)-2-hydroxypropionate

A155456-100G A155456-25G
77877-028EA 14 USD
77877-028 77877-029
Butyl (S)-(-)-lactate ≥97%
Butyl (S)-(-)-lactate

Precaución: for Research Use Only

Formula: C₇H₁₄O₃
MDL Number: MFCD00004519
CAS Number: 34451-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR