Methyl 4,5-Dibromo-2-furancarboxylate ≥95%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dibromo-2-furancarboxylic acid methyl ester, 4,5-Dibromo-2-furoic acid methyl ester, Methyl 4,5-Dibromo-2-furoate

A155425-5G A155425-100MG A155425-1G A155425-250MG
77592-910EA 129.15 USD
77592-910 77881-217 77881-218 77881-219
Methyl 4,5-Dibromo-2-furancarboxylate ≥95%
Methyl 4,5-Dibromo-2-furancarboxylate

Precaución: for Research Use Only

Formula: C₆H₄Br₂O₃
MDL Number: MFCD00092493
CAS Number: 54113-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR