4-((S)-3-Acryloyl-8-chloro-10-fluoro-12-oxo-2,3,4,4a,5,6-hexahydro-1H,12H-benzo[b]pyrazino[1,2-e][1,5]oxazocin-9-yl)-2-amino-7-fluorobenzo[b]thiophene-3-carbonitrile ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
KRAS G12C inhibitor 19
A1553137-1MG
A1553137-10MG
A1553137-5MG
77598-062EA
100.45
USD
77598-062
77625-412
77625-414
4-((S)-3-Acryloyl-8-chloro-10-fluoro-12-oxo-2,3,4,4a,5,6-hexahydro-1H,12H-benzo[b]pyrazino[1,2-e][1,5]oxazocin-9-yl)-2-amino-7-fluorobenzo[b]thiophene-3-carbonitrile ≥98%
4-((S)-3-Acryloyl-8-chloro-10-fluoro-12-oxo-2,3,4,4a,5,6-hexahydro-1H,12H-benzo[b]pyrazino[1,2-e][1,5]oxazocin-9-yl)-2-amino-7-fluorobenzo[b]thiophene-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₂₅H₁₉ClF₂N₄O₃S MW: 528.96 g/mol |
CAS Number:
2649788-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...