6-(2-Methoxypyrimidin-5-yl)benzo[d]thiazol-2-amine ≥98%

Supplier: Ambeed
Warning

Synonyms: FTO-IN-3

A1552517-10MG A1552517-1MG A1552517-5MG
77598-056EA 492 USD
77598-056 77598-058 77598-060
6-(2-Methoxypyrimidin-5-yl)benzo[d]thiazol-2-amine ≥98%
6-(2-Methoxypyrimidin-5-yl)benzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₄OS
MW: 258.3 g/mol
MDL Number: MFCD34602595
CAS Number: 2585198-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR