(2,5-Dihydroxyphenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1550937-1G A1550937-250MG
77876-292EA 271 USD
77876-292 77876-293
(2,5-Dihydroxyphenyl)boronic acid ≥95%
(2,5-Dihydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BO₄
MW: 153.93 g/mol
MDL Number: MFCD23381021
CAS Number: 1238196-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR