(R)-2-Methyl-3-(1-((4-methyl-7-morpholinopyrido[3,4-d]pyridazin-1-yl)amino)ethyl)benzonitrile ≥95%

Supplier: Ambeed
Warning

A1550907-100MG A1550907-25MG A1550907-50MG
77876-289EA 485 USD
77876-289 77876-290 77876-291
(R)-2-Methyl-3-(1-((4-methyl-7-morpholinopyrido[3,4-d]pyridazin-1-yl)amino)ethyl)benzonitrile ≥95%
(R)-2-Methyl-3-(1-((4-methyl-7-morpholinopyrido[3,4-d]pyridazin-1-yl)amino)ethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₆O
CAS Number: 2654743-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR