2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl 4-methylbenzenesulfonate ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Tos-PEG2-NH-Boc
A155045-250MG
A155045-5G
A155045-1G
77746-064EA
19
USD
77746-064
77746-065
77746-063
2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl 4-methylbenzenesulfonate ≥98%
2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₆H₂₅NO₆S MW: 359.43 g/mol |
MDL Number:
MFCD28506265 CAS Number: 192132-77-7 UN: 2810 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...