(R)-1-((7-Cyano-2-(3'-((3-(((R)-3-hydroxypyrrolidin-1-yl)methyl)-1,7-naphthyridin-8-yl)amino)-2,2'-dimethyl-[1,1'-biphenyl]-3-yl)benzo[d]oxazol-5-yl)methyl)pyrrolidine-3-carboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: INCB086550

A1550238-10MG A1550238-1MG A1550238-5MG
77598-034EA 136.33 USD
77598-034 77598-036 77598-038
(R)-1-((7-Cyano-2-(3'-((3-(((R)-3-hydroxypyrrolidin-1-yl)methyl)-1,7-naphthyridin-8-yl)amino)-2,2'-dimethyl-[1,1'-biphenyl]-3-yl)benzo[d]oxazol-5-yl)methyl)pyrrolidine-3-carboxylic acid ≥98%
(R)-1-((7-Cyano-2-(3'-((3-(((R)-3-hydroxypyrrolidin-1-yl)methyl)-1,7-naphthyridin-8-yl)amino)-2,2'-dimethyl-[1,1'-biphenyl]-3-yl)benzo[d]oxazol-5-yl)methyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₄₁H₃₉N₇O₄
MW: 693.79 g/mol
MDL Number: MFCD34549780
CAS Number: 2230911-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR