(R)-N-(5-((4-((5-Chloro-4-fluoro-2-(2-hydroxypropan-2-yl)phenyl)amino)pyrimidin-2-yl)amino)-2-(3-(dimethylamino)pyrrolidin-1-yl)-4-methoxyphenyl)acrylamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Sunvozertinib

A1550231-1MG A1550231-10MG A1550231-5MG
77598-032EA 18.45 USD
77598-032 77625-400 77625-402
(R)-N-(5-((4-((5-Chloro-4-fluoro-2-(2-hydroxypropan-2-yl)phenyl)amino)pyrimidin-2-yl)amino)-2-(3-(dimethylamino)pyrrolidin-1-yl)-4-methoxyphenyl)acrylamide ≥98%
(R)-N-(5-((4-((5-Chloro-4-fluoro-2-(2-hydroxypropan-2-yl)phenyl)amino)pyrimidin-2-yl)amino)-2-(3-(dimethylamino)pyrrolidin-1-yl)-4-methoxyphenyl)acrylamide

Precaución: for Research Use Only

Formula: C₂₉H₃₅ClFN₇O₃
MW: 584.08 g/mol
MDL Number: MFCD34567621
CAS Number: 2370013-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR