N-(4-(1-Methyl-1H-pyrazol-4-yl)benzyl)-6-(7-(3-(pyrrolidin-1-yl)propoxy)imidazo[1,2-a]pyridin-3-yl)pyrimidin-4-amine ≥98%

Supplier: Ambeed
Warning

Synonyms: M4205

A1550120-10MG A1550120-1MG A1550120-5MG
77598-024EA 369 USD
77598-024 77598-026 77598-028
N-(4-(1-Methyl-1H-pyrazol-4-yl)benzyl)-6-(7-(3-(pyrrolidin-1-yl)propoxy)imidazo[1,2-a]pyridin-3-yl)pyrimidin-4-amine ≥98%
N-(4-(1-Methyl-1H-pyrazol-4-yl)benzyl)-6-(7-(3-(pyrrolidin-1-yl)propoxy)imidazo[1,2-a]pyridin-3-yl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₂₉H₃₂N₈O
MW: 508.62 g/mol
MDL Number: MFCD34368512
CAS Number: 2590556-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR