2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester ≥97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 2,9-diazaspiro[5.5]undecane-9-carboxylate, 9-Boc-2,9-diazaspiro[5.5]undecane

A154872-5G A154872-1G
77600-204EA 184.5 USD
77600-204 77667-190
2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester ≥97%
2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₄H₂₆N₂O₂
MDL Number: MFCD10700117
CAS Number: 1023595-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR