2-Bromo-4-methoxy-1,3-dimethylbenzene ≥95%

Supplier: Ambeed
Warning

A1548215-1G A1548215-250MG
77592-900EA 130.18 USD
77592-900 77625-382
2-Bromo-4-methoxy-1,3-dimethylbenzene ≥95%
2-Bromo-4-methoxy-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
MDL Number: MFCD28401429
CAS Number: 23453-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR