N-(2-((3S,4R)-3-Fluoro-4-methoxypiperidin-1-yl)pyrimidin-4-yl)-5-isopropyl-8-((2R,3S)-2-methyl-3-((methylsulfonyl)methyl)azetidin-1-yl)isoquinolin-3-amine ≥95%

Supplier: Ambeed
Warning

Synonyms: BLU-945

A1547532-1MG A1547532-10MG A1547532-5MG A1547532-100MG A1547532-250MG A1547532-25MG A1547532-50MG
77598-020EA 120.95 USD
77598-020 77625-376 77625-378 77876-256 77876-257 77876-258 77876-259
N-(2-((3S,4R)-3-Fluoro-4-methoxypiperidin-1-yl)pyrimidin-4-yl)-5-isopropyl-8-((2R,3S)-2-methyl-3-((methylsulfonyl)methyl)azetidin-1-yl)isoquinolin-3-amine ≥95%
N-(2-((3S,4R)-3-Fluoro-4-methoxypiperidin-1-yl)pyrimidin-4-yl)-5-isopropyl-8-((2R,3S)-2-methyl-3-((methylsulfonyl)methyl)azetidin-1-yl)isoquinolin-3-amine

Precaución: for Research Use Only

Formula: C₂₈H₃₇FN₆O₃S
MW: 556.7 g/mol
CAS Number: 2660250-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR