(R)-1-(3-Methoxyphenyl)propan-1-amine hydrochloride ≥95%

Supplier: Ambeed
Warning

A154712-1G A154712-100MG A154712-250MG
77671-182EA 211 USD
77671-182 77881-210 77881-211
(R)-1-(3-Methoxyphenyl)propan-1-amine hydrochloride ≥95%
(R)-1-(3-Methoxyphenyl)propan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆ClNO
MDL Number: MFCD12757163
CAS Number: 856562-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR