6-(Trifluoromethoxy)-4-quinolinol ≥97%

Supplier: Ambeed
Warning

A154642-1G A154642-5G
77879-068EA 13 USD
77879-068 77879-069
6-(Trifluoromethoxy)-4-quinolinol ≥97%
6-(Trifluoromethoxy)-4-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO₂
MDL Number: MFCD00219858
CAS Number: 175203-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR