1-(3-Bromophenyl)cyclopropanecarbonitrile ≥95%

Supplier: Ambeed
Warning

A154623-5G A154623-25G
77885-264EA 47 USD
77885-264 77885-263
1-(3-Bromophenyl)cyclopropanecarbonitrile ≥95%
1-(3-Bromophenyl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MDL Number: MFCD07374414
CAS Number: 124276-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR