Methyl-6-bromo-1-benzofuran-2-carboxylate ≥95%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-bromobenzofuran-2-carboxylate

A154572-1G A154572-250MG A154572-5G
77611-468EA 272.95 USD
77611-468 77683-296 77683-297
Methyl-6-bromo-1-benzofuran-2-carboxylate ≥95%
Methyl-6-bromo-1-benzofuran-2-carboxylate

Precaución: For research use only.

Formula: C₁₀H₇BrO₃
MW: 255.07 g/mol
MDL Number: MFCD22126097
CAS Number: 425675-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR