(5S,5'S)-5,5'-(((((2,2'-Dichloro-[1,1'-biphenyl]-3,3'-diyl)bis(3-methoxypyrazine-5,2-diyl))bis(methylene))bis(azanediyl))bis(methylene))bis(pyrrolidin-2-one) ≥95%

Supplier: Ambeed
Warning

A1545172-10MG A1545172-1MG A1545172-5MG
77598-012EA 159.9 USD
77598-012 77598-014 77598-016
(5S,5'S)-5,5'-(((((2,2'-Dichloro-[1,1'-biphenyl]-3,3'-diyl)bis(3-methoxypyrazine-5,2-diyl))bis(methylene))bis(azanediyl))bis(methylene))bis(pyrrolidin-2-one) ≥95%
(5S,5'S)-5,5'-(((((2,2'-Dichloro-[1,1'-biphenyl]-3,3'-diyl)bis(3-methoxypyrazine-5,2-diyl))bis(methylene))bis(azanediyl))bis(methylene))bis(pyrrolidin-2-one)

Precaución: for Research Use Only

Formula: C₃₄H₃₆Cl₂N₈O₄
MDL Number: MFCD33402309
CAS Number: 2374856-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR